APO455826939 Apollo Scientific Ltd

Ethyl 5-(bromoacetyl)isoxazole-3-carboxylate

  • MDL number MFCD00173773
  • Cas Number : 104776-74-1
  • Molecular Weight : 262.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-73
Synonym Ethyl 5-(bromoacetyl)-1,2-oxazole-3-carboxylate, 5-(Bromoacetyl)-3-(ethoxycarbonyl)isoxazole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-bromoacetylisoxazole-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-bromoacetylisoxazole-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart