APO455826951 Apollo Scientific Ltd

2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride

  • MDL number MFCD08445576
  • Cas Number : 31788-96-2
  • Molecular Weight : 263.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-phenyl-2-(1-pyrrolidinyl)ethanamine dihydrochloride
Inchi Code 1S/C12H18N2.2ClH/c13-10-12(14-8-4-5-9-14)11-6-2-1-3-7-11;;/h1-3,6-7,12H,4-5,8-10,13H2;2*1H
Inchi Key HXIUHWWUUPRKDB-UHFFFAOYSA-N

Description

Synonym 1-(2-Amino-1-phenylethyl)pyrrolidine dihydrochloride, beta-(Pyrrolidin-1-yl)phenethylamine dihydrochloride
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-phenyl-2-pyrrolidin-1-ylethylamine-dihydrochloride

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/2-phenyl-2-pyrrolidin-1-ylethylamine-dihydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart