APO455826952 Apollo Scientific Ltd

N1,N1-Dimethyl-2-phenylethane-1,2-diamine

  • MDL number MFCD01862528
  • Cas Number : 31788-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym beta-Amino-N,N-dimethylphenethylamine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/n1n1-dimethyl-2-phenylethane-12-diamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/n1n1-dimethyl-2-phenylethane-12-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart