APO455826960 Apollo Scientific Ltd

2-(4-Chlorophenyl)-2-phenylethylamine hydrochloride

  • MDL number MFCD02089402
  • Cas Number : 21998-50-5
  • Molecular Weight : 268.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-chlorophenyl)-2-phenylethanamine hydrochloride
Inchi Code 1S/C14H14ClN.ClH/c15-13-8-6-12(7-9-13)14(10-16)11-4-2-1-3-5-11;/h1-9,14H,10,16H2;1H
Inchi Key KYGYPNIZCSHFEW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-chlorophenyl-2-phenylethylamine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-4-chlorophenyl-2-phenylethylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart