APO455826965 Apollo Scientific Ltd

2-Amino-6-bromobenzoic acid

  • MDL number MFCD03618455
  • Cas Number : 20776-48-1
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-137
Synonym 3-Bromo-2-carboxyaniline, 6-Bromoanthranilic acid
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-bromobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-bromobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart