APO455826972 Apollo Scientific Ltd

4-(4-Methoxyphenoxy)piperidine hydrochloride

  • MDL number MFCD03840124
  • Cas Number : 333954-89-5
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-4-methoxyphenoxypiperidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methoxyphenoxypiperidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart