APO455826975 Apollo Scientific Ltd

4-(2-Methoxyphenoxy)piperidine hydrochloride

  • MDL number MFCD03840131
  • Cas Number : 950649-22-6
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(Piperidin-4-yl)oxy]anisole hydrochloride, 1-Methoxy-2-[(piperidin-4-yl)oxy]benzene hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-2-methoxyphenoxypiperidine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P261
MSDS https://store.apolloscientific.co.uk/product/4-2-methoxyphenoxypiperidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart