APO455826977 Apollo Scientific Ltd

2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride

  • MDL number MFCD03840122
  • Cas Number : 950649-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(Piperidin-4-yl)oxy]-1,4-diazine dihydrochloride, 4-[(Pyrazin-2-yl)oxy]piperidine dihydrochloride
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27964

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27964
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart