APO455826978 Apollo Scientific Ltd

3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride

  • MDL number MFCD00084916
  • Cas Number : 160842-62-6
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-tert-butyl-1-methyl-1H-pyrazole-5-carbonyl chloride
Inchi Code 1S/C9H13ClN2O/c1-9(2,3)7-5-6(8(10)13)12(4)11-7/h5H,1-4H3
Inchi Key SJGURXGUNLVMAQ-UHFFFAOYSA-N

Description

Synonym 3-(tert-Butyl)-5-(chlorocarbonyl)-1-methyl-1H-pyrazole, 3-(tert-Butyl)-5-(chloroformyl)-1-methyl-1H-pyrazole
Flash Point 117/15mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-tert-butyl-1-methyl-1h-pyrazole-5-carbonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-tert-butyl-1-methyl-1h-pyrazole-5-carbonyl-chloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart