APO455826980 Apollo Scientific Ltd

3-(2-Amino-1-hydroxyethyl)pyridine dihydrochloride

  • MDL number MFCD09028161
  • Cas Number : 101012-01-5
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-1-(pyridin-3-yl)ethan-1-ol dihydrochloride, 2-Hydroxy-2-(pyridin-3-yl)ethylamine dihydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-2-amino-1-hydroxyethylpyridine-dihydrochloride

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-2-amino-1-hydroxyethylpyridine-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart