APO455826984 Apollo Scientific Ltd

3-(4-Chlorophenyl)morpholine hydrochloride

  • MDL number MFCD03840089
  • Cas Number : 1170797-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-4-(morpholin-3-yl)benzene hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-4-chlorophenylmorpholine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-chlorophenylmorpholine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart