APO455826987 Apollo Scientific Ltd

3-(2-Chlorophenyl)morpholine hydrochloride

  • MDL number MFCD03840094
  • Cas Number : 1172507-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2-(morpholin-3-yl)benzene hydrochloride
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28001

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=28001
Availability details for
Quantity
  • Availability
  • Add to Cart