APO455826990 Apollo Scientific Ltd

4,4-Diphenylpiperidine hydrochloride

  • MDL number MFCD01700799
  • Cas Number : 63675-71-8
  • Molecular Weight : 273.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/44-diphenylpiperidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/44-diphenylpiperidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart