APO455826994 Apollo Scientific Ltd

2-[(Piperidin-4-yl)methyl]pyridine dihydrochloride

  • MDL number MFCD03840134
  • Cas Number : 886886-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(Pyridin-2-yl)methyl]piperidine dihydrochloride
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-piperidin-4-ylmethylpyridine-dihydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-piperidin-4-ylmethylpyridine-dihydrochloride
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart