APO455826997 Apollo Scientific Ltd

2-(Pyridin-3-yl)piperazine trihydrochloride

  • MDL number MFCD09702426
  • Cas Number : 1171887-03-8
  • Molecular Weight : 272.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Piperazin-2-yl)pyridine trihydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-pyridin-3-ylpiperazine-trihydrochloride

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-pyridin-3-ylpiperazine-trihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart