APO455826998 Apollo Scientific Ltd

3-Phenylpiperazine, N1-BOC protected

  • MDL number MFCD03840082
  • Cas Number : 502649-25-4
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-phenylpiperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-phenylpiperazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenylpiperazine-n1-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenylpiperazine-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart