APO455827002 Apollo Scientific Ltd

2-[(Pyrazin-2-yl)oxy]benzaldehyde

  • MDL number MFCD03840028
  • Cas Number : 887344-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2-Formylphenoxy)pyrazine, 2-(2-Formylphenoxy)-1,4-diazine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-pyrazin-2-yloxybenzaldehyde

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-pyrazin-2-yloxybenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart