APO455827003 Apollo Scientific Ltd

3-[(Pyrazin-2-yl)oxy]benzaldehyde

  • MDL number MFCD03840111
  • Cas Number : 887344-44-7
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-pyrazinyloxy)benzaldehyde
Inchi Code 1S/C11H8N2O2/c14-8-9-2-1-3-10(6-9)15-11-7-12-4-5-13-11/h1-8H
Inchi Key XBVLRDWQVHJLHH-UHFFFAOYSA-N

Description

Synonym 2-(3-Formylphenoxy)pyrazine, 2-(3-Formylphenoxy)-1,4-diazine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-pyrazin-2-yloxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-pyrazin-2-yloxybenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart