APO455827007 Apollo Scientific Ltd

3-[(Pyridin-3-yl)methyl]piperidine-3-carboxylic acid, N-BOC protected

  • MDL number MFCD03840045
  • Cas Number : 887344-18-5
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butoxycarbonyl)-3-(pyridin-3-ylmethyl)piperidine-3-carboxylic acid
Inchi Code 1S/C17H24N2O4/c1-16(2,3)23-15(22)19-9-5-7-17(12-19,14(20)21)10-13-6-4-8-18-11-13/h4,6,8,11H,5,7,9-10,12H2,1-3H3,(H,20,21)
Inchi Key LNKSCFFYJGBNAU-UHFFFAOYSA-N

Description

Synonym 1-(tert-Butoxycarbonyl)-3-[(pyridin-3-yl)methyl]piperidine-3-carboxylic acid, 1-(tert-Butoxycarbonyl)-3-carboxy-3-[(pyridin-3-yl)methyl]piperidine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-pyridin-3-ylmethylpiperidine-3-carboxylic-acid-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-pyridin-3-ylmethylpiperidine-3-carboxylic-acid-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart