APO455827012 Apollo Scientific Ltd

3,4-Dihydro-6-hydroxynaphthalen-2(1H)-one

  • MDL number MFCD05663283
  • Cas Number : 52727-28-3
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-146
Synonym 6-Hydroxy-2-tetralone, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydronaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 125/3mm(sub.)
Product Link https://store.apolloscientific.co.uk/product/34-dihydro-6-hydroxynaphthalen-21h-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/34-dihydro-6-hydroxynaphthalen-21h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart