APO455827019 Apollo Scientific Ltd

1-(2-Amino-3-methylbut-1-yl)-1H-imidazole dihydrochloride

  • MDL number MFCD04117727
  • Cas Number : 1172472-37-5
  • Molecular Weight : 226.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(1H-Imidazol-1-yl)-1-isopropylethylamine dihydrochloride, 1-(1H-Imidazol-1-yl)-3-methylbutan-2-amine dihydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-2-amino-3-methylbut-1-yl-1h-imidazole-dihydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-amino-3-methylbut-1-yl-1h-imidazole-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart