APO455827020 Apollo Scientific Ltd

4-Hydroxy-4-(pyridin-3-yl)piperidine

  • MDL number MFCD04117758
  • Cas Number : 50461-59-1
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Pyridin-3-yl)piperidin-4-ol
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-4-pyridin-3-ylpiperidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-4-pyridin-3-ylpiperidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart