APO455827021 Apollo Scientific Ltd

2-(Bromoacetyl)pyridine hydrobromide

  • MDL number MFCD02181196
  • Cas Number : 17570-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Synonym 2-Bromo-1-(pyridin-2-yl)ethan-1-one hydrobromide, 2-(2-Bromoethanoyl)pyridine hydrobromide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13148

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13148
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart