APO455827053 Apollo Scientific Ltd

5-Bromopyridine-2-carboxamide

  • MDL number MFCD04066715
  • Cas Number : 90145-48-5
  • Molecular Weight : 201.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Synonym 5-Bromopicolinamide, 5-Bromo-2-carbamoylpyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromopyridine-2-carboxamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromopyridine-2-carboxamide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart