APO455827061 Apollo Scientific Ltd

Bis(4-bromophenyl)acetylene

  • MDL number MFCD00185147
  • Cas Number : 2789-89-1
  • Molecular Weight : 336.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-187
Synonym 1,1'-(Ethyne-1,2-diyl)bis(4-bromobenzene)
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/bis4-bromophenylacetylene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/bis4-bromophenylacetylene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart