APO455827064 Apollo Scientific Ltd

4-Chloro-3-nitrobenzeneboronic acid

  • MDL number MFCD02258950
  • Cas Number : 151169-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 275
Synonym 5-Borono-2-chloronitrobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-nitrobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-nitrobenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart