APO455827067 Apollo Scientific Ltd

4-Chloro-2-methoxybenzeneboronic acid

  • MDL number MFCD02684317
  • Cas Number : 762287-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-134
Synonym 2-Borono-5-chloroanisole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart