APO455827068 Apollo Scientific Ltd

(Cyclopent-1-en-1-yl)boronic acid

  • MDL number MFCD02179496
  • Cas Number : 850036-28-1
  • Molecular Weight : 111.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 134-140
Iupac Name 1-cyclopenten-1-ylboronic acid
Inchi Code 1S/C5H9BO2/c7-6(8)5-3-1-2-4-5/h3,7-8H,1-2,4H2
Inchi Key UZBHNSVUMGIKLU-UHFFFAOYSA-N

Description

Synonym 1-Boronocyclopent-1-ene
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 134-140
Product Link https://store.apolloscientific.co.uk/product/cyclopent-1-en-1-ylboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/cyclopent-1-en-1-ylboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart