APO455827074 Apollo Scientific Ltd

3-(Phenylcarbamoyl)benzeneboronic acid

  • MDL number MFCD04115711
  • Cas Number : 397843-71-9
  • Molecular Weight : 241.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-260
Synonym 3-[(Phenylamino)carbonyl]benzeneboronic acid, 3-Borono-N-phenylbenzamide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart