APO455827083 Apollo Scientific Ltd

1-(Pyridin-4-yl)thiourea

  • MDL number MFCD00068177
  • Cas Number : 164670-44-4
  • Molecular Weight : 153.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-184
Synonym 4-(Carbamothioylamino)pyridine, 4-Thioureidopyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-pyridin-4-ylthiourea

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-pyridin-4-ylthiourea
pictograms GHS06
hstatements H301;H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart