APO455827086 Apollo Scientific Ltd

1,3-Thiazole-4-carboxaldehyde

  • MDL number MFCD00626896
  • Cas Number : 3364-80-5
  • Molecular Weight : 113.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-65
Synonym 4-Formyl-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/13-thiazole-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-thiazole-4-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart