APO455827096 Apollo Scientific Ltd

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD03011525
  • Cas Number : 81282-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-163
Synonym 1-(4-Carboxyphenyl)-3,5-dimethyl-1H-pyrazole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28298

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28298
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart