APO455827107 Apollo Scientific Ltd

2-Bromo-5-chloroanisole

  • MDL number MFCD03790889
  • Cas Number : 174913-09-8
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-chloroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-chloroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart