APO455827112 Apollo Scientific Ltd

3-Chloropyridine-4-boronic acid, neopentyl glycol ester

  • MDL number MFCD06796314
  • Cas Number : 915070-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine, 2-(3-Chloropyridin-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28330

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28330
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart