APO455827113 Apollo Scientific Ltd

4-Chloromandelic acid

  • MDL number MFCD00042724
  • Cas Number : 492-86-4
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-122
Synonym (4-Chlorophenyl)(hydroxy)acetic acid, 2-(4-Chlorophenyl)-2-hydroxyethanoic acid
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloromandelic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloromandelic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart