APO455827115 Apollo Scientific Ltd

2-(Dimethylamino)benzaldehyde

  • MDL number MFCD03407377
  • Cas Number : 579-72-6
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(dimethylamino)benzaldehyde
Inchi Code 1S/C9H11NO/c1-10(2)9-6-4-3-5-8(9)7-11/h3-7H,1-2H3
Inchi Key DGPBVJWCIDNDPN-UHFFFAOYSA-N

Description

Synonym N,N-Dimethyl-2-formylaniline, N,N-Dimethylanthranilaldehyde
Purity 97%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-dimethylaminobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-dimethylaminobenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart