APO455827122 Apollo Scientific Ltd

3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde

  • MDL number MFCD00058678
  • Cas Number : 178686-24-3
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Ethoxy-4-formyl-6-nitrophenol
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-ethoxy-4-hydroxy-5-nitrobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-ethoxy-4-hydroxy-5-nitrobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart