APO455827123 Apollo Scientific Ltd

4-[(Methylamino)sulphonyl]benzene boronic acid

  • MDL number MFCD06659864
  • Cas Number : 226396-31-2
  • Molecular Weight : 215.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-140
Synonym [4-(Methylsulfamoyl)phenyl]boronic acid, Methyl 4-boronobenzenesulfonamide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylaminosulphonylbenzene-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylaminosulphonylbenzene-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart