APO455827126 Apollo Scientific Ltd

1,2-Dihydro-1-methyl-2-oxoquinoline-4-carboxaldehyde

  • MDL number MFCD03942175
  • Cas Number : 15112-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/12-dihydro-1-methyl-2-oxoquinoline-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/12-dihydro-1-methyl-2-oxoquinoline-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart