APO455827127 Apollo Scientific Ltd

4-[(Methylsulphonylamino)methyl]benzeneboronic acid

  • MDL number MFCD06659816
  • Cas Number : 850568-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-239
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29428

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=29428
Availability details for
Quantity
  • Availability
  • Add to Cart