APO455827130 Apollo Scientific Ltd

Ethyl 4-amino-3-carbamoylisothiazole-5-carboxylate

  • MDL number MFCD00848452
  • Cas Number : 54968-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 4-amino-3-(aminocarbonyl)isothiazole-5-carboxylate, 4-Amino-3-carbamoyl-5-(ethoxycarbonyl)isothiazole, 4-Amino-3-carbamoyl-5-(ethoxycarbonyl)-1,2-thiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38623

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38623
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart