APO455827133 Apollo Scientific Ltd

3-Amino-4-chlorobenzeneboronic acid, pinacol ester

  • MDL number MFCD06659926
  • Cas Number : 850567-56-5
  • Molecular Weight : 253.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Synonym 6-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-chlorobenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-chlorobenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart