APO455827134 Apollo Scientific Ltd

5-Amino-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD02641259
  • Cas Number : 400073-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239
Synonym 5-Amino-4-carboxy-2-phenyl-2H-1,2,3-triazole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38847

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38847
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart