APO455827135 Apollo Scientific Ltd

4-Amino-1,2,5-oxadiazole-3-carbonitrile

  • MDL number MFCD00235169
  • Cas Number : 156463-85-3
  • Molecular Weight : 110.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Aminofurazan-3-carbonitrile, 3-Amino-4-cyano-1,2,5-oxadiazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-125-oxadiazole-3-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-125-oxadiazole-3-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart