APO455827136 Apollo Scientific Ltd

5-Amino-3-(cyanomethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00022387
  • Cas Number : 54711-21-6
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-200
Synonym 5-Amino-3-(cyanomethyl)-1H-pyrazol-4-yl cyanide, (5-Amino-4-cyano-1H-pyrazol-3-yl)acetonitrile
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-cyanomethyl-1h-pyrazole-4-carbonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-cyanomethyl-1h-pyrazole-4-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart