APO455827140 Apollo Scientific Ltd

5-Amino-3-(2-aminoethyl)-1H-1,2,4-triazole dihydrochloride

  • MDL number MFCD00460101
  • Cas Number : 99839-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(2-Aminoethyl)-1H-1,2,4-triazol-5-amine dihydrochloride, 2-(5-Amino-1H-1,2,4-triazol-3-yl)ethylamine dihydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-2-aminoethyl-1h-124-triazole-dihydrochloride

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-2-aminoethyl-1h-124-triazole-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart