APO455827146 Apollo Scientific Ltd

5,7-Dimethylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD00461394
  • Cas Number : 300691-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-5,7-dimethylpyrazolo[1,5-a]pyrimidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/57-dimethylpyrazolo15-apyrimidine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/57-dimethylpyrazolo15-apyrimidine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart