APO455827147 Apollo Scientific Ltd

4-Amino-3-phenylbutanoic acid hydrochloride

  • MDL number MFCD00075860
  • Cas Number : 3060-41-1
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-201
Synonym beta-(Carboxymethyl)phenethylamine hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-phenylbutanoic-acid-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-phenylbutanoic-acid-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart