APO455827153 Apollo Scientific Ltd

Ethyl 6-amino-5-bromonicotinate

  • MDL number MFCD06659894
  • Cas Number : 850429-51-5
  • Molecular Weight : 245.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-127
Synonym Ethyl 2-amino-3-bromo-5-pyridinecarboxylate
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-6-amino-5-bromonicotinate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-6-amino-5-bromonicotinate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart