APO455827160 Apollo Scientific Ltd

3-Nitro-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00195039
  • Cas Number : 39205-87-3
  • Molecular Weight : 138.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Nitro-1H-pyrazol-4-yl cyanide, 4-Cyano-3-nitro-1H-pyrazole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-nitro-1h-pyrazole-4-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-nitro-1h-pyrazole-4-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart